missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium Acetate 95.0+%, TCI America™

$171.49 - $544.30

Chemical Identifiers

CAS 143314-17-4
Molecular Formula C8H14N2O2
Molecular Weight (g/mol) 170.212
MDL Number MFCD06798186
InChI Key XIYUIMLQTKODPS-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium acetate, basionic tm bc 01, emim ac, 1-ethyl-3-methylimidazoliumacetate, basionics™ bc 01, acmc-1c26d, 1h-imidazolium, 3-ethyl-1-methyl-, acetate 1:1, 1-methyl-3-ethylimidazolium acetate, 1-ethyl-3-methylimidazolium acetate hplc
PubChem CID 11658353
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;acetate
SMILES CCN1C=C[N+](=C1)C.CC(=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E08835G
View Documents
TCI America
E08835G
5 g
Each for $171.49
Only null left
 
E088325G
View Documents
TCI America
E088325G
25 g
Each for $544.30
Only null left
 
Description

Description

Specifications

Chemical Identifiers

143314-17-4
170.212
XIYUIMLQTKODPS-UHFFFAOYSA-M
11658353
CCN1C=C[N+](=C1)C.CC(=O)[O-]
C8H14N2O2
MFCD06798186
1-ethyl-3-methylimidazolium acetate, basionic tm bc 01, emim ac, 1-ethyl-3-methylimidazoliumacetate, basionics™ bc 01, acmc-1c26d, 1h-imidazolium, 3-ethyl-1-methyl-, acetate 1:1, 1-methyl-3-ethylimidazolium acetate, 1-ethyl-3-methylimidazolium acetate hplc
1-ethyl-3-methylimidazol-3-ium;acetate

Specifications

143314-17-4
MFCD06798186
1-ethyl-3-methylimidazolium acetate, basionic tm bc 01, emim ac, 1-ethyl-3-methylimidazoliumacetate, basionics™ bc 01, acmc-1c26d, 1h-imidazolium, 3-ethyl-1-methyl-, acetate 1:1, 1-methyl-3-ethylimidazolium acetate, 1-ethyl-3-methylimidazolium acetate hplc
CCN1C=C[N+](=C1)C.CC(=O)[O-]
170.212
170.21
Liquid
C8H14N2O2
5 g
XIYUIMLQTKODPS-UHFFFAOYSA-M
1-ethyl-3-methylimidazol-3-ium;acetate
11658353
≥95.0% (HPLC,T)
1-Ethyl-3-methylimidazolium Acetate
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'