missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Ethyl-3-methylimidazolium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
$150.10 - $531.27
Chemical Identifiers
CAS | 174899-82-2 |
---|---|
Molecular Formula | C8H11F6N3O4S2 |
Molecular Weight (g/mol) | 391.303 |
MDL Number | MFCD03788927 |
InChI Key | LRESCJAINPKJTO-UHFFFAOYSA-N |
PubChem CID | 11731903 |
IUPAC Name | bis(trifluoromethylsulfonyl)azanide;1-ethyl-3-methylimidazol-3-ium |
SMILES | CCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F |
Chemical Identifiers
174899-82-2 | |
391.303 | |
LRESCJAINPKJTO-UHFFFAOYSA-N | |
bis(trifluoromethylsulfonyl)azanide;1-ethyl-3-methylimidazol-3-ium |
C8H11F6N3O4S2 | |
MFCD03788927 | |
11731903 | |
CCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F |
Specifications
174899-82-2 | |
Yellow | |
MFCD03788927 | |
LRESCJAINPKJTO-UHFFFAOYSA-N | |
bis(trifluoromethylsulfonyl)azanide;1-ethyl-3-methylimidazol-3-ium | |
11731903 | |
≥98.0% (HPLC,T) | |
1-Ethyl-3-methylimidazolium Bis(trifluoromethanesulfonyl)imide |
-15°C | |
C8H11F6N3O4S2 | |
5 g | |
CCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F | |
391.303 | |
391.30 | |
Liquid |
Safety and Handling
TSCA : No