missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethyl-3-methylimidazolium ethyl sulfate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2757118

 View more versions of this product

Catalog No. AAH2757118


Explore available promotions

Description

Description

1-Ethyl-3-methylimidazolium ethyl sulfate is an ionic liquid having free form chloride used as an alternative to the corresponding halide salts in metathesis reactions to prepare other ionic liquids including 1-butyl-3-methylimidazolium hexafluorophosphate. It is also utilized in the study of layering and shear properties of ionic liquid between nano-films and mica surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethyl-3-methylimidazolium ethyl sulfate is an ionic liquid having free form chloride used as an alternative to the corresponding halide salts in metathesis reactions to prepare other ionic liquids including 1-butyl-3-methylimidazolium hexafluorophosphate. It is also utilized in the study of layering and shear properties of ionic liquid between nano-films and mica surfaces.

Solubility
Miscible with water and alcohol.

Notes
Hygroscopic. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and bases.
Specifications

Specifications

1-Ethyl-3-methylimidazolium ethyl sulfate
<-30°C
162°C (324°F)
MFCD06798189
1-ethyl-3-methylimidazolium ethyl sulfate, 1-ethyl-3-methylimidazolium ethylsulfate, 1-ethyl-3-methyl-1h-imidazol-3-ium ethyl sulfate, basionics™ lq 01, dsstox_cid_29130, dsstox_rid_83349, dsstox_gsid_49274, ksc221o0f, 1h-imidazolium, 3-ethyl-1-methyl-, ethyl sulfate 1:1, 1-ethyl-3-me-thylimidazolium ethylsulfate
VRFOKYHDLYBVAL-UHFFFAOYSA-M
1-ethyl-3-methylimidazol-3-ium;ethyl sulfate
12095229
99%
342573-75-5
1.24
C8H16N2O4S
50 g
Miscible with water and alcohol.
CCN1C=C[N+](=C1)C.CCOS(=O)(=O)[O-]
236.286
236.29

Chemical Identifiers

342573-75-5
236.286
VRFOKYHDLYBVAL-UHFFFAOYSA-M
12095229
CCN1C=C[N+](=C1)C.CCOS(=O)(=O)[O-]
C8H16N2O4S
MFCD06798189
1-ethyl-3-methylimidazolium ethyl sulfate, 1-ethyl-3-methylimidazolium ethylsulfate, 1-ethyl-3-methyl-1h-imidazol-3-ium ethyl sulfate, basionics™ lq 01, dsstox_cid_29130, dsstox_rid_83349, dsstox_gsid_49274, ksc221o0f, 1h-imidazolium, 3-ethyl-1-methyl-, ethyl sulfate 1:1, 1-ethyl-3-me-thylimidazolium ethylsulfate
1-ethyl-3-methylimidazol-3-ium;ethyl sulfate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only