missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Ethyl-3-methylimidazolium methyl sulfate, 98%, Thermo Scientific Chemicals
$65.37 - $381.67
Chemical Identifiers
CAS | 516474-01-4 |
---|---|
Molecular Formula | C7H14N2O4S |
Molecular Weight (g/mol) | 222.259 |
MDL Number | MFCD08457697 |
InChI Key | BXSDLSWVIAITRQ-UHFFFAOYSA-M |
Synonym | 1-ethyl-3-methylimidazolium methyl sulfate, 1-ethyl-3-methylimidazolium methylsulfate, 1-ethyl-3-methyl-1h-imidazol-3-ium methyl sulfate, 1-ethyl-3-methylimidazolium methyl sulfate 1-, 1-ethyl-3-methylimidazoliummethylsulfate, dsstox_cid_29266, dsstox_rid_83385, dsstox_gsid_49310, 1-ethyl-3-methylimidazolium methylsul-fate, 1-ethyl-3-methylimidazolium methyl sulfate hplc |
PubChem CID | 16211409 |
IUPAC Name | 1-ethyl-3-methylimidazol-3-ium;methyl sulfate |
SMILES | CCN1C=C[N+](=C1)C.COS(=O)(=O)[O-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5948006
|
Thermo Scientific Chemicals
H5948006 |
5 g |
Each for $65.37
|
|
|||||
AAH5948018
|
Thermo Scientific Chemicals
H5948018 |
50 g |
Each for $381.67
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
516474-01-4 | |
222.259 | |
BXSDLSWVIAITRQ-UHFFFAOYSA-M | |
16211409 | |
CCN1C=C[N+](=C1)C.COS(=O)(=O)[O-] |
C7H14N2O4S | |
MFCD08457697 | |
1-ethyl-3-methylimidazolium methyl sulfate, 1-ethyl-3-methylimidazolium methylsulfate, 1-ethyl-3-methyl-1h-imidazol-3-ium methyl sulfate, 1-ethyl-3-methylimidazolium methyl sulfate 1-, 1-ethyl-3-methylimidazoliummethylsulfate, dsstox_cid_29266, dsstox_rid_83385, dsstox_gsid_49310, 1-ethyl-3-methylimidazolium methylsul-fate, 1-ethyl-3-methylimidazolium methyl sulfate hplc | |
1-ethyl-3-methylimidazol-3-ium;methyl sulfate |
Specifications
516474-01-4 | |
MFCD08457697 | |
Moisture sensitive | |
BXSDLSWVIAITRQ-UHFFFAOYSA-M | |
1-ethyl-3-methylimidazol-3-ium;methyl sulfate | |
16211409 | |
98% |
C7H14N2O4S | |
5 g | |
1-ethyl-3-methylimidazolium methyl sulfate, 1-ethyl-3-methylimidazolium methylsulfate, 1-ethyl-3-methyl-1h-imidazol-3-ium methyl sulfate, 1-ethyl-3-methylimidazolium methyl sulfate 1-, 1-ethyl-3-methylimidazoliummethylsulfate, dsstox_cid_29266, dsstox_rid_83385, dsstox_gsid_49310, 1-ethyl-3-methylimidazolium methylsul-fate, 1-ethyl-3-methylimidazolium methyl sulfate hplc | |
CCN1C=C[N+](=C1)C.COS(=O)(=O)[O-] | |
222.259 | |
222.26 | |
1-Ethyl-3-methylimidazolium methyl sulfate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only