missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethylcyclohexanol, 97%, Thermo Scientific™

Chemical Identifiers

CAS 1940-18-7
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00021402
InChI Key BUCJHJXFXUZJHL-UHFFFAOYSA-N
Synonym 1-ethylcyclohexanol, cyclohexanol, 1-ethyl, unii-1ves5dbc5c, 1-aethyl-cyclohexanol-1, 1-ethyl-1-cyclohexanol, 1ves5dbc5c, 1-aethyl-cyclohexanol-1 german, ethylcyclohexanol, cyclohexanol, ethyl, 1-ethyl-cyclohexan-1-ol
PubChem CID 16021
IUPAC Name 1-ethylcyclohexan-1-ol
SMILES CCC1(O)CCCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC453350050
SDS View Documents
Thermo Scientific Chemicals
453350050
5mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC453350250
SDS View Documents
Thermo Scientific Chemicals
453350250
25mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1940-18-7
128.22
BUCJHJXFXUZJHL-UHFFFAOYSA-N
16021
CCC1(O)CCCCC1
C8H16O
MFCD00021402
1-ethylcyclohexanol, cyclohexanol, 1-ethyl, unii-1ves5dbc5c, 1-aethyl-cyclohexanol-1, 1-ethyl-1-cyclohexanol, 1ves5dbc5c, 1-aethyl-cyclohexanol-1 german, ethylcyclohexanol, cyclohexanol, ethyl, 1-ethyl-cyclohexan-1-ol
1-ethylcyclohexan-1-ol

Specifications

1940-18-7
166.0°C
96% min. (GC)
MFCD00021402
0.919
BUCJHJXFXUZJHL-UHFFFAOYSA-N
1-ethylcyclohexan-1-ol
16021
97%
0.9190g/mL
68°C
C8H16O
5mL
1-ethylcyclohexanol, cyclohexanol, 1-ethyl, unii-1ves5dbc5c, 1-aethyl-cyclohexanol-1, 1-ethyl-1-cyclohexanol, 1ves5dbc5c, 1-aethyl-cyclohexanol-1 german, ethylcyclohexanol, cyclohexanol, ethyl, 1-ethyl-cyclohexan-1-ol
CCC1(O)CCCCC1
128.22
128.22
1-Ethylcyclohexanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only