Learn More
1-Ethylcyclopentanol, 96%, Thermo Scientific Chemicals
$231.26 - $231.26
Chemical Identifiers
| CAS | 1462-96-0 |
|---|---|
| Molecular Formula | C7H14O |
| Molecular Weight (g/mol) | 114.188 |
| MDL Number | MFCD00019281 |
| InChI Key | LPCWIFPJLFCXRS-UHFFFAOYSA-N |
| Synonym | 1-ethylcyclopentanol, cyclopentanol, 1-ethyl, 1-ethyl-1-cyclopentanol, acmc-1bzot, ksc181q7f, 1-ethyl-1-hydroxycyclopentane |
| PubChem CID | 137018 |
| IUPAC Name | 1-ethylcyclopentan-1-ol |
| SMILES | CCC1(CCCC1)O |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
AAH6680906
|
Thermo Scientific Chemicals
H6680906 |
5 g |
Each for $231.26
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1462-96-0 | |
| 114.188 | |
| LPCWIFPJLFCXRS-UHFFFAOYSA-N | |
| 137018 | |
| CCC1(CCCC1)O |
| C7H14O | |
| MFCD00019281 | |
| 1-ethylcyclopentanol, cyclopentanol, 1-ethyl, 1-ethyl-1-cyclopentanol, acmc-1bzot, ksc181q7f, 1-ethyl-1-hydroxycyclopentane | |
| 1-ethylcyclopentan-1-ol |
Specifications
| 1462-96-0 | |
| 0.909 g/mL | |
| C7H14O | |
| MFCD00019281 | |
| UN1987 | |
| 1-ethylcyclopentanol, cyclopentanol, 1-ethyl, 1-ethyl-1-cyclopentanol, acmc-1bzot, ksc181q7f, 1-ethyl-1-hydroxycyclopentane | |
| CCC1(CCCC1)O | |
| 114.188 | |
| 114.19 | |
| Liquid |
| Colorless to Yellow | |
| 49°C (120°F) | |
| 1.457 | |
| 5 g | |
| Moisture sensitive | |
| LPCWIFPJLFCXRS-UHFFFAOYSA-N | |
| 1-ethylcyclopentan-1-ol | |
| 137018 | |
| 96% | |
| 1-Ethylcyclopentanol |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only