missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethylpiperazine-2,3-dione, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1824818

 View more versions of this product

Catalog No. AAA1824818


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Ethylpiperazine-2,3-dione
113°C to 124°C
MFCD00051825
879076
ZBEKOEYCWKIMGU-UHFFFAOYSA-N
1-ethylpiperazine-2,3-dione
108812
97%
59702-31-7
C6H10N2O2
50 g
1-ethyl-2,3-dioxopiperazine, n-ethyl-2,3-dioxopiperazine, 1-ethyl-2,3-piperazinedione, 2,3-piperazinedione, 1-ethyl, 4-ethyl-2,3-dioxopiperazine, 4-ethyl-2,3-piperazinedione, ccris 8931, 4-ethyl-2,3-dioxo-piperazine, 4-ethyl-2,3-dioxypiperazine, n-ethylpiperazine-2,3-dione
CCN1CCNC(=O)C1=O
142.158
142.16

Chemical Identifiers

59702-31-7
142.158
ZBEKOEYCWKIMGU-UHFFFAOYSA-N
108812
CCN1CCNC(=O)C1=O
C6H10N2O2
MFCD00051825
1-ethyl-2,3-dioxopiperazine, n-ethyl-2,3-dioxopiperazine, 1-ethyl-2,3-piperazinedione, 2,3-piperazinedione, 1-ethyl, 4-ethyl-2,3-dioxopiperazine, 4-ethyl-2,3-piperazinedione, ccris 8931, 4-ethyl-2,3-dioxo-piperazine, 4-ethyl-2,3-dioxypiperazine, n-ethylpiperazine-2,3-dione
1-ethylpiperazine-2,3-dione
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 261-866-5

RTECSNumber : TL6380250

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only