Learn More
1-Ethylpiperidine, 99%
CAS: 766-09-6 | C7H15N | 113.2 g/mol
$212.87 - $864.34
Chemical Identifiers
| CAS | 766-09-6 |
|---|---|
| Molecular Formula | C7H15N |
| Molecular Weight (g/mol) | 113.2 |
| MDL Number | MFCD00006507 |
| InChI Key | HTLZVHNRZJPSMI-UHFFFAOYSA-N |
| Synonym | n-ethylpiperidine, piperidine, 1-ethyl, n-aethylpiperidin, n-ethyl piperidine, ethylpiperidine, n-aethylpiperidin german, 1-ethyl-piperidine, n-ethyl-piperidine, 1-ethyl piperidine, 1-ethylpiperidene |
| PubChem CID | 13007 |
| ChEBI | CHEBI:39017 |
| IUPAC Name | 1-ethylpiperidine |
| SMILES | CCN1CCCCC1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC118885000
|
Thermo Scientific Chemicals
118885000 |
500 mL | Glass bottle |
Each for $212.87
|
|
||||
|
AC118880025
|
Thermo Scientific Chemicals
118880025 |
2.5 L | Glass bottle |
Each for $864.34
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 766-09-6 | |
| 113.2 | |
| HTLZVHNRZJPSMI-UHFFFAOYSA-N | |
| 13007 | |
| 1-ethylpiperidine |
| C7H15N | |
| MFCD00006507 | |
| n-ethylpiperidine, piperidine, 1-ethyl, n-aethylpiperidin, n-ethyl piperidine, ethylpiperidine, n-aethylpiperidin german, 1-ethyl-piperidine, n-ethyl-piperidine, 1-ethyl piperidine, 1-ethylpiperidene | |
| CHEBI:39017 | |
| CCN1CCCCC1 |
Specifications
| 766-09-6 | |
| 100.0 | |
| Colorless to Yellow | |
| 129.0°C to 131.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4430 to 1.4450 | |
| 500 mL | |
| 0.824 | |
| Solubility in water: 50g/L (20°C) | |
| CCN1CCCCC1 | |
| 113.2 | |
| CHEBI:39017 | |
| 99% | |
| 1-Ethylpiperidine, 99% |
| 98.5 | |
| -20.0°C | |
| 0.8240g/mL | |
| 17°C | |
| 98.5% min. (GC) | |
| C7H15N | |
| MFCD00006507 | |
| 20,17 | |
| n-ethylpiperidine, piperidine, 1-ethyl, n-aethylpiperidin, n-ethyl piperidine, ethylpiperidine, n-aethylpiperidin german, 1-ethyl-piperidine, n-ethyl-piperidine, 1-ethyl piperidine, 1-ethylpiperidene | |
| HTLZVHNRZJPSMI-UHFFFAOYSA-N | |
| 1-ethylpiperidine | |
| 13007 | |
| 113.2 | |
| Liquid |
Safety and Handling
GHS H Statement
Toxic if inhaled.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Highly flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger
EINECSNumber : 212-161-6
RTECSNumber : TN0250000
TSCA : TSCA
RUO – Research Use Only