missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethylpyrrole 98.0+%, TCI America™
$41.02 - $156.59
Chemical Identifiers
| CAS | 617-92-5 |
|---|---|
| Molecular Formula | C6H9N |
| Molecular Weight (g/mol) | 95.15 |
| MDL Number | MFCD00060633 |
| InChI Key | VPUAYOJTHRDUTK-UHFFFAOYSA-N |
| PubChem CID | 185226 |
| IUPAC Name | 1-ethyl-1H-pyrrole |
| SMILES | CCN1C=CC=C1 |