missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Ethynyl-1-cyclohexanol 98.0+%, TCI America™
Supplier: TCI America E0297500G
Specifications
1-Ethynyl-1-cyclohexanol | |
White-Yellow | |
C8H12O | |
500 g | |
1-ethynyl-1-cyclohexanol, 1-ethynylcyclohexanol, ethynylcyclohexanol, cyclohexanol, 1-ethynyl, 1-hydroxycyclohexyl ethyne, 1-ethynyl-1-hydroxycyclohexane, 1-ethynyl cyclohexanol, 1-ethynyl-cyclohexanol, unii-6rv04025eh, 1-ethynyl-cyclohexan-1-ol | |
C#CC1(CCCCC1)O | |
124.183 | |
124.18 | |
Crystalline Powder |
78-27-3 | |
174°C | |
MFCD00003858 | |
2811 | |
QYLFHLNFIHBCPR-UHFFFAOYSA-N | |
1-ethynylcyclohexan-1-ol | |
6525 | |
≥98.0% (GC) |
Chemical Identifiers
78-27-3 | |
124.183 | |
QYLFHLNFIHBCPR-UHFFFAOYSA-N | |
6525 | |
C#CC1(CCCCC1)O |
C8H12O | |
MFCD00003858 | |
1-ethynyl-1-cyclohexanol, 1-ethynylcyclohexanol, ethynylcyclohexanol, cyclohexanol, 1-ethynyl, 1-hydroxycyclohexyl ethyne, 1-ethynyl-1-hydroxycyclohexane, 1-ethynyl cyclohexanol, 1-ethynyl-cyclohexanol, unii-6rv04025eh, 1-ethynyl-cyclohexan-1-ol | |
1-ethynylcyclohexan-1-ol |
Safety and Handling
EINECSNumber : (3)-3256
RTECSNumber : GV9100000
TSCA : Yes