missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Ethynyl-4-(trifluoromethoxy)benzene 97.0+%, TCI America™
$225.14 - $675.44
Chemical Identifiers
CAS | 160542-02-9 |
---|---|
Molecular Formula | C9H5F3O |
Molecular Weight (g/mol) | 186.13 |
MDL Number | MFCD03094333 |
InChI Key | RWWGGRCLMVYXPM-UHFFFAOYSA-N |
Synonym | 4-(Trifluoromethoxy)phenylacetylene |
PubChem CID | 2778494 |
IUPAC Name | 1-ethynyl-4-(trifluoromethoxy)benzene |
SMILES | FC(F)(F)OC1=CC=C(C=C1)C#C |
Chemical Identifiers
160542-02-9 | |
186.13 | |
RWWGGRCLMVYXPM-UHFFFAOYSA-N | |
2778494 | |
FC(F)(F)OC1=CC=C(C=C1)C#C |
C9H5F3O | |
MFCD03094333 | |
4-(Trifluoromethoxy)phenylacetylene | |
1-ethynyl-4-(trifluoromethoxy)benzene |
Specifications
160542-02-9 | |
C9H5F3O | |
1 g | |
4-(Trifluoromethoxy)phenylacetylene | |
FC(F)(F)OC1=CC=C(C=C1)C#C | |
186.13 | |
186.13 | |
Liquid |
Yellow | |
MFCD03094333 | |
1993 | |
RWWGGRCLMVYXPM-UHFFFAOYSA-N | |
1-ethynyl-4-(trifluoromethoxy)benzene | |
2778494 | |
≥97.0% (GC) | |
1-Ethynyl-4-(trifluoromethoxy)benzene |
Safety and Handling
TSCA : No