missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Fluoro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1187114

 View more versions of this product

Catalog No. AAA1187114

Explore available promotions

Description

Description

1-Fluoro-2,4-dinitrobenzene is used to identify the amino acid sequence. It reacts with amino group of amino acids to yield dinitrophenyl-amino acids. It is also used in chromatographic methods. Further, it acts as an alkylating agent used in elucidating amino acid sequence in proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Fluoro-2,4-dinitrobenzene is used to identify the amino acid sequence. It reacts with amino group of amino acids to yield dinitrophenyl-amino acids. It is also used in chromatographic methods. Further, it acts as an alkylating agent used in elucidating amino acid sequence in proteins.

Solubility
Soluble in chloroform, benzene, ether and propylene glycol.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

1-Fluoro-2,4-dinitrobenzene
24°C to 30°C
296°C
C6H3FN2O4
MFCD00007056
UN2811
14,4172
Soluble in chloroform,benzene,ether and propylene glycol.
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])F
186.098
CHEBI:53049
99%
70-34-8
1.482
>110°C (230°F)
1.569
25 g
398632
2,4-dinitrofluorobenzene, dinitrofluorobenzene, fdnb, dnfb, 2,4-dinitro-1-fluorobenzene, sanger's reagent, fluoro-2,4-dinitrobenzene, 2,4-dinitrobenzenefluoride, 2,4-dinitrophenyl fluoride, fluorodinitrobenzene
LOTKRQAVGJMPNV-UHFFFAOYSA-N
1-fluoro-2,4-dinitrobenzene
6264
186.1

Chemical Identifiers

70-34-8
186.098
LOTKRQAVGJMPNV-UHFFFAOYSA-N
6264
1-fluoro-2,4-dinitrobenzene
C6H3FN2O4
MFCD00007056
2,4-dinitrofluorobenzene, dinitrofluorobenzene, fdnb, dnfb, 2,4-dinitro-1-fluorobenzene, sanger's reagent, fluoro-2,4-dinitrobenzene, 2,4-dinitrobenzenefluoride, 2,4-dinitrophenyl fluoride, fluorodinitrobenzene
CHEBI:53049
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])F
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H318-H341-H315-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye damage.
Suspected of causing genetic defects.
Causes skin irritation.
May cause an allergic skin reaction.

P260-P262-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P314-P330-P333+P313-P362-P501c

H301+H311+H331-H315-H317-H320-H335-H336-H373

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 200-734-3

RTECSNumber : CZ7800000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only