missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Heptanol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 120360025

 View more versions of this product

Catalog No. AC120360025

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Heptanol
0.5mg KOH/g max.
100.0
0.8219g/mL
73°C
97.5% min. (GC)
C7H16O
CH3(CH2)6OH
2.5 L
0.01% max.
15, 4696
Solubility in water: 2.85g/L (100°C). Other solubilities: 5.15g/L water (130°C), soluble in ethanol, ether and usual organic, solvents
CCCCCCCO
116.20
116.2
Extra Pure
111-70-6
97.5
-35.0°C
176.0°C
Authentic
Glass bottle
1.4230 to 1.4250
MFCD00002986
01, 414
0.8219
1-heptanol, heptyl alcohol, heptanol, n-heptanol, n-heptyl alcohol, enanthic alcohol, gentanol, 1-hydroxyheptane, n-heptan-1-ol, enanthyl alcohol
BBMCTIGTTCKYKF-UHFFFAOYSA-N
heptan-1-ol
8129
98%
Liquid

Chemical Identifiers

111-70-6
116.20
BBMCTIGTTCKYKF-UHFFFAOYSA-N
8129
CCCCCCCO
C7H16O
MFCD00002986
1-heptanol, heptyl alcohol, heptanol, n-heptanol, n-heptyl alcohol, enanthic alcohol, gentanol, 1-hydroxyheptane, n-heptan-1-ol, enanthyl alcohol
heptan-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 203-897-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only