missing translation for 'onlineSavingsMsg'
Learn More

1-Heptanol, 99%

CAS: 111-70-6 | C7H16O | 116.20 g/mol

Supplier:  Thermo Scientific Chemicals A12793AP

 View more versions of this product

Catalog No. AAA12793AP

Only null left
Explore available promotions

Description

Description

1-Heptanol, is used in the fragrance industry due to its pleasant smell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Heptanol, is used in the fragrance industry due to its pleasant smell.

Solubility
Soluble in water 1.7 g/L at 25°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Stable under recommended storage conditions. Incompatible to strong oxidizing agents, acid chlorides, acid anhydrides.
Specifications

Specifications

1-Heptanol
0.820
C7H16O
MFCD00002986
1731686
1-heptanol, heptyl alcohol, heptanol, n-heptanol, n-heptyl alcohol, enanthic alcohol, gentanol, 1-hydroxyheptane, n-heptan-1-ol, enanthyl alcohol
CCCCCCCO
116.20
116.2
Liquid
111-70-6
99%
CH3(CH2)6OH
500 mL
14,4661
BBMCTIGTTCKYKF-UHFFFAOYSA-N
heptan-1-ol
8129
99%

Chemical Identifiers

111-70-6
116.20
BBMCTIGTTCKYKF-UHFFFAOYSA-N
8129
CCCCCCCO
C7H16O
MFCD00002986
1-heptanol, heptyl alcohol, heptanol, n-heptanol, n-heptyl alcohol, enanthic alcohol, gentanol, 1-hydroxyheptane, n-heptan-1-ol, enanthyl alcohol
heptan-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H319-H227

GHS P Statement
P210-P280-P305+P351+P338-P301+P312-P312-P363-P322-P337+P313-P403+P235-P501a

Warning

RTECSNumber : MK0350000

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only