missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Hexanethiol, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 215275000

 View more versions of this product

Catalog No. AC215275000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Hexanethiol
95.0
-81°C to 80°C
0.8400g/mL
30°C
95% min. (GC)
C6H14S
CH3(CH2)5SH
500 mL
0.84
Solubility in water: insoluble. Other solubilities: soluble in alcohols, ethyl ether, hydrocarbons
CCCCCCS
118.24
118.24
Liquid
111-31-9
100.0
Colorless to Yellow
150.0°C to 154.0°C
Authentic
Glass bottle
1.4470 to 1.4510
MFCD00004909
01, 408
1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan
PMBXCGGQNSVESQ-UHFFFAOYSA-N
hexane-1-thiol
8106
96%

Chemical Identifiers

111-31-9
118.24
PMBXCGGQNSVESQ-UHFFFAOYSA-N
8106
CCCCCCS
C6H14S
MFCD00004909
1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan
hexane-1-thiol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED

GHS Signal Word: Danger

EINECSNumber : 203-857-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only