Learn More
1-Hexene, 97%, Thermo Scientific Chemicals
1-Hexene, 97%, C6H12, CAS Number-592-41-6, dialene 6, butyl ethylene, 1-hexene, butylethylene, 1-n-hexene, hexylene, hexene-1, hexene, n-hexene, unii-b38zz8c206, 1L, 96% min. (GC), 01, 215, 62.0 deg.C to 65.0 deg.C, 100.0, 96.0, CHEBI:24579, Colorless, 0.6780g/mL, 209-753-1, -26 deg.C, 84.15
$59.78 - $197.82
Chemical Identifiers
CAS | 592-41-6 |
---|---|
Molecular Formula | C6H12 |
Molecular Weight (g/mol) | 84.15 |
MDL Number | MFCD00009505 |
InChI Key | LIKMAJRDDDTEIG-UHFFFAOYSA-N |
Synonym | 1-hexene, hexene, butylethylene, hexylene, hexene-1, 1-n-hexene, butyl ethylene, dialene 6, n-hexene, unii-b38zz8c206 |
PubChem CID | 11597 |
ChEBI | CHEBI:24579 |
IUPAC Name | hex-1-ene |
SMILES | CCCCC=C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC213211000
|
Thermo Scientific Chemicals
213211000 |
100 mL | Glass bottle |
Each for $59.78
|
|
|||||
AC213210010
|
Thermo Scientific Chemicals
213210010 |
1 L | Glass bottle |
Each for $85.17
|
|
|||||
AC213210025
|
Thermo Scientific Chemicals
213210025 |
2.5 L | Glass bottle |
Each for $197.82
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
592-41-6 | |
84.15 | |
LIKMAJRDDDTEIG-UHFFFAOYSA-N | |
11597 | |
hex-1-ene |
C6H12 | |
MFCD00009505 | |
1-hexene, hexene, butylethylene, hexylene, hexene-1, 1-n-hexene, butyl ethylene, dialene 6, n-hexene, unii-b38zz8c206 | |
CHEBI:24579 | |
CCCCC=C |
Specifications
592-41-6 | |
100.0 | |
Colorless | |
62.0°C to 65.0°C | |
Authentic | |
Glass bottle | |
1.3867 to 1.3887 | |
MFCD00009505 | |
01, 215 | |
1-hexene, hexene, butylethylene, hexylene, hexene-1, 1-n-hexene, butyl ethylene, dialene 6, n-hexene, unii-b38zz8c206 | |
LIKMAJRDDDTEIG-UHFFFAOYSA-N | |
hex-1-ene | |
11597 | |
CHEBI:24579 | |
97% | |
1-Hexene |
96.0 | |
-140.0°C | |
0.6780g/mL | |
−26°C | |
96% min. (GC) | |
C6H12 | |
CH3(CH2)3CH=CH2 | |
100 mL | |
0.678 | |
Solubility in water: 50 mg/l (20°C). Other solubilities: soluble in alcohol and hydrocarbon solvents, soluble in ethanol and diethyl ether | |
CCCCC=C | |
84.15 | |
0.34 mm2/s (40°C) | |
84.15 | |
Liquid |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take precautionary measures against static discharge.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Imm
GHS Signal Word: Danger
EINECSNumber : 209-753-1
RUO – Research Use Only