missing translation for 'onlineSavingsMsg'
Learn More

1-Hexyl-3-methylimidazolium Bromide 98.0+%, TCI America™

$233.00 - $699.02

Chemical Identifiers

CAS 85100-78-3
Molecular Formula C10H19BrN2
Molecular Weight (g/mol) 247.18
MDL Number MFCD03427612
InChI Key BGSUDDILQRFOKZ-UHFFFAOYSA-M
PubChem CID 2734237
IUPAC Name 1-hexyl-3-methylimidazol-3-ium;bromide
SMILES CCCCCCN1C=C[N+](=C1)C.[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H12275G
View Documents
TCI America
H12275G
5 g
Each for $233.00
Only null left
 
H122725G
View Documents
TCI America
H122725G
25 g
Each for $699.02
Only null left
 
Description

Description

Specifications

Chemical Identifiers

85100-78-3
247.18
BGSUDDILQRFOKZ-UHFFFAOYSA-M
1-hexyl-3-methylimidazol-3-ium;bromide
C10H19BrN2
MFCD03427612
2734237
CCCCCCN1C=C[N+](=C1)C.[Br-]

Specifications

85100-78-3
C10H19BrN2
5 g
CCCCCCN1C=C[N+](=C1)C.[Br-]
247.18
247.18
Liquid
395°C
MFCD03427612
BGSUDDILQRFOKZ-UHFFFAOYSA-M
1-hexyl-3-methylimidazol-3-ium;bromide
2734237
≥98.0% (HPLC,T)
1-Hexyl-3-methylimidazolium Bromide
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'