missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Hexylamine, 99%, Thermo Scientific Chemicals

$136.22 - $447.41

Chemical Identifiers

CAS 111-26-2
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00008240
InChI Key BMVXCPBXGZKUPN-UHFFFAOYSA-N
Synonym hexylamine, 1-aminohexane, n-hexylamine, 1-hexanamine, 1-hexylamine, mono-n-hexylamine, hexanamine, hexyl amine, unii-ci4e002zv8, hexyl-amine
PubChem CID 8102
ChEBI CHEBI:5712
IUPAC Name hexan-1-amine
SMILES CCCCCCN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA15663AK
View Documents
Thermo Scientific Chemicals
A15663AK
250 mL
Each for $136.22
 
AAA15663AU
View Documents
Thermo Scientific Chemicals
A15663AU
1000 mL
Each for $447.41
 
Description

Description

1-Hexylamine is used as corrosion inhibitors, emulsifiers and surfactants. It is used in dyes, rubber and pharmaceutical industries. It is also used to study the modification of alkanethlol monolayers in gas-phase surface reactions. It acts as an initiator in the ring opening polymerization of N-carboxyanhydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexylamine is used as corrosion inhibitors, emulsifiers and surfactants. It is used in dyes, rubber and pharmaceutical industries. It is also used to study the modification of alkanethlol monolayers in gas-phase surface reactions. It acts as an initiator in the ring opening polymerization of N-carboxyanhydride.

Solubility
Miscible with water, dichloromethane, alcohol and acetone.

Notes
Air sensitive. Incompatible with acids, acid chlorides, carbon dioxide and strong oxidizing agents.
Specifications

Chemical Identifiers

111-26-2
101.19
BMVXCPBXGZKUPN-UHFFFAOYSA-N
8102
hexan-1-amine
C6H15N
MFCD00008240
hexylamine, 1-aminohexane, n-hexylamine, 1-hexanamine, 1-hexylamine, mono-n-hexylamine, hexanamine, hexyl amine, unii-ci4e002zv8, hexyl-amine
CHEBI:5712
CCCCCCN

Specifications

111-26-2
0.773
129°C to 131°C
Amine-like
1.418
MFCD00008240
UN2734
Air Sensitive
Miscible with water,dichloromethane,alcohol and acetone.
CCCCCCN
101.19
CHEBI:5712
99%
-19°C
11.6
27°C (80°F)
C6H15N
CH3(CH2)5NH2
250 mL
1731298
hexylamine, 1-aminohexane, n-hexylamine, 1-hexanamine, 1-hexylamine, mono-n-hexylamine, hexanamine, hexyl amine, unii-ci4e002zv8, hexyl-amine
BMVXCPBXGZKUPN-UHFFFAOYSA-N
hexan-1-amine
8102
101.19
1-Hexylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H301+H311-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.

EINECSNumber : 203-851-8

RTECSNumber : MQ4540000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only