missing translation for 'onlineSavingsMsg'
Learn More

1-Hexyne, 98%

CAS: 693-02-7 | C6H10 | 82.13 g/mol

$97.60 - $259.86

Chemical Identifiers

CAS 693-02-7
Molecular Formula C6H10
Molecular Weight (g/mol) 82.13
InChI Key CGHIBGNXEGJPQZ-UHFFFAOYSA-N
Synonym 1-hexyne, n-butylacetylene, butylacetylene, hexyne, unii-5fzf2f38f5, n-hexyne, 1-hexyn, hex-5-yn-1-yl, pubchem13057
PubChem CID 12732
IUPAC Name hex-1-yne
SMILES CCCCC#C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC329900250
SDS View Documents
Thermo Scientific Chemicals
329900250
25 mL Glass bottle
Each for $97.60
Only null left
 
AC329901000
SDS View Documents
Thermo Scientific Chemicals
329901000
100 mL Glass bottle
Each for $259.86
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

693-02-7
82.13
1-hexyne, n-butylacetylene, butylacetylene, hexyne, unii-5fzf2f38f5, n-hexyne, 1-hexyn, hex-5-yn-1-yl, pubchem13057
hex-1-yne
C6H10
CGHIBGNXEGJPQZ-UHFFFAOYSA-N
12732
CCCCC#C

Specifications

693-02-7
Colorless
71.0°C to 72.0°C
Authentic
Glass bottle
1.3974
25 mL
0.71
CGHIBGNXEGJPQZ-UHFFFAOYSA-N
hex-1-yne
12732
98%
1-Hexyne
-132.0°C
0.7100g/mL
−021°C
97.5% min. (GC)
C6H10
CH3(CH2)3C≡CH
01,253
1-hexyne, n-butylacetylene, butylacetylene, hexyne, unii-5fzf2f38f5, n-hexyne, 1-hexyn, hex-5-yn-1-yl, pubchem13057
CCCCC#C
82.13
82.13
Liquid
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 211-736-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only