missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxy-2-naphthaldehyde 98.0+%, TCI America™

$210.63 - $631.92

Chemical Identifiers

CAS 574-96-9
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.183
MDL Number MFCD00092325
InChI Key OITQDWKMIPXGFL-UHFFFAOYSA-N
Synonym 1-Naphthol-2-carboxaldehyde
PubChem CID 443195
ChEBI CHEBI:28091
IUPAC Name 1-hydroxynaphthalene-2-carbaldehyde
SMILES C1=CC=C2C(=C1)C=CC(=C2O)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H08391G
View Documents
TCI America
H08391G
1 g
Each for $210.63
Only null left
 
H08395G
View Documents
TCI America
H08395G
5 g
Each for $631.92
Only null left
 
Description

Description

Specifications

Chemical Identifiers

574-96-9
172.183
OITQDWKMIPXGFL-UHFFFAOYSA-N
443195
1-hydroxynaphthalene-2-carbaldehyde
C11H8O2
MFCD00092325
1-Naphthol-2-carboxaldehyde
CHEBI:28091
C1=CC=C2C(=C1)C=CC(=C2O)C=O

Specifications

574-96-9
Green-Yellow
MFCD00092325
1-Naphthol-2-carboxaldehyde
C1=CC=C2C(=C1)C=CC(=C2O)C=O
172.183
CHEBI:28091
≥98.0% (GC)
1-Hydroxy-2-naphthaldehyde
56°C
C11H8O2
1 g
OITQDWKMIPXGFL-UHFFFAOYSA-N
1-hydroxynaphthalene-2-carbaldehyde
443195
172.18
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'