missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Hydroxybenzotriazole Monohydrate 97.0+%, TCI America™

Supplier:  TCI America H0468200G

 View more versions of this product

Catalog No. H0468200G


Explore available promotions

Description

Description

Specifications

Specifications

1-Hydroxybenzotriazole Monohydrate
160°C
C6H7N3O2
200 g
1,2,3-Benzotriazol-1-ol, HOBt
O.ON1N=NC2=CC=CC=C12
153.14
135.13
Crystalline Powder
80029-43-2
White
MFCD00065690
3474
PJUPKRYGDFTMTM-UHFFFAOYSA-N
1H-1,2,3-benzotriazol-1-ol hydrate
2796029
≥97.0% (T)

Chemical Identifiers

80029-43-2
153.14
PJUPKRYGDFTMTM-UHFFFAOYSA-N
2796029
O.ON1N=NC2=CC=CC=C12
C6H7N3O2
MFCD00065690
1,2,3-Benzotriazol-1-ol, HOBt
1H-1,2,3-benzotriazol-1-ol hydrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-3579&(5)-3603

RTECSNumber : DM1288000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions