missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1-Hydroxycyclohexyl)(4-methoxyphenyl)acetonitrile 98.0+%, TCI America™
$174.48 - $174.48
Chemical Identifiers
| CAS | 93413-76-4 |
|---|---|
| Molecular Formula | C15H19NO2 |
| Molecular Weight (g/mol) | 245.322 |
| MDL Number | MFCD06658142 |
| InChI Key | ASYJSBPNAIDUHX-UHFFFAOYSA-N |
| Synonym | 1-[Cyano(4-methoxyphenyl)methyl]cyclohexanol |
| PubChem CID | 9899621 |
| IUPAC Name | 2-(1-hydroxycyclohexyl)-2-(4-methoxyphenyl)acetonitrile |
| SMILES | COC1=CC=C(C=C1)C(C#N)C2(CCCCC2)O |
Chemical Identifiers
| 93413-76-4 | |
| 245.322 | |
| ASYJSBPNAIDUHX-UHFFFAOYSA-N | |
| 9899621 | |
| COC1=CC=C(C=C1)C(C#N)C2(CCCCC2)O |
| C15H19NO2 | |
| MFCD06658142 | |
| 1-[Cyano(4-methoxyphenyl)methyl]cyclohexanol | |
| 2-(1-hydroxycyclohexyl)-2-(4-methoxyphenyl)acetonitrile |
Specifications
| 93413-76-4 | |
| White | |
| MFCD06658142 | |
| 1-[Cyano(4-methoxyphenyl)methyl]cyclohexanol | |
| COC1=CC=C(C=C1)C(C#N)C2(CCCCC2)O | |
| 245.322 | |
| 245.32 | |
| Crystalline Powder |
| 127°C | |
| C15H19NO2 | |
| 1 g | |
| ASYJSBPNAIDUHX-UHFFFAOYSA-N | |
| 2-(1-hydroxycyclohexyl)-2-(4-methoxyphenyl)acetonitrile | |
| 9899621 | |
| ≥98.0% (HPLC,N) | |
| (1-Hydroxycyclohexyl)(4-methoxyphenyl)acetonitrile |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator