Learn More
1-Hydroxycyclohexyl phenyl ketone, 98%, Thermo Scientific Chemicals
$77.99 - $191.75
Chemical Identifiers
CAS | 947-19-3 |
---|---|
Molecular Formula | C13H16O2 |
Molecular Weight (g/mol) | 204.269 |
MDL Number | MFCD00059561 |
InChI Key | QNODIIQQMGDSEF-UHFFFAOYSA-N |
Synonym | 1-hydroxycyclohexyl phenyl ketone, 1-hydroxycyclohexyl phenyl methanone, 1-benzoylcyclohexanol, hydroxycyclohexyl phenyl ketone, 1-benzoylcyclohexan-1-ol, irgacure 184, methanone, 1-hydroxycyclohexyl phenyl, unii-e7jvn2243x, 1-hydroxycyclohexyl-phenylmethanone, 1-cyclohexylhydroxyphenylketone |
PubChem CID | 70355 |
IUPAC Name | (1-hydroxycyclohexyl)-phenylmethanone |
SMILES | C1CCC(CC1)(C(=O)C2=CC=CC=C2)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL1137706
|
Thermo Scientific Chemicals
L1137706 |
5 g |
Each for $77.99
|
|
||||||
AAL1137714
|
Thermo Scientific Chemicals
L1137714 |
25 g |
Each for $191.75
|
|
||||||
Description
1-Hydroxycyclohexyl phenyl ketone is used as photoinitiator in UV-radiation-curable technologies which are used in various applications and industry branches such as printing and packaging, coatings, furniture and flooring and adhesives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Hydroxycyclohexyl phenyl ketone is used as photoinitiator in UV-radiation-curable technologies which are used in various applications and industry branches such as printing and packaging, coatings, furniture and flooring and adhesives.
Solubility
Slightly soluble in water (1108 mg/L at 25°C). Soluble in acetone, butyl acetate, methanol and toluene.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
947-19-3 | |
204.269 | |
QNODIIQQMGDSEF-UHFFFAOYSA-N | |
70355 | |
C1CCC(CC1)(C(=O)C2=CC=CC=C2)O |
C13H16O2 | |
MFCD00059561 | |
1-hydroxycyclohexyl phenyl ketone, 1-hydroxycyclohexyl phenyl methanone, 1-benzoylcyclohexanol, hydroxycyclohexyl phenyl ketone, 1-benzoylcyclohexan-1-ol, irgacure 184, methanone, 1-hydroxycyclohexyl phenyl, unii-e7jvn2243x, 1-hydroxycyclohexyl-phenylmethanone, 1-cyclohexylhydroxyphenylketone | |
(1-hydroxycyclohexyl)-phenylmethanone |
Specifications
947-19-3 | |
1.17 | |
>150°C (302°F) | |
MFCD00059561 | |
2330973 | |
Slightly soluble in water (1108mg/L at 25°C). Soluble in acetone,butyl acetate,methanol and toluene. | |
C1CCC(CC1)(C(=O)C2=CC=CC=C2)O | |
204.269 | |
204.27 | |
1-Hydroxycyclohexyl phenyl ketone |
46°C to 49°C | |
>225°C | |
C13H16O2 | |
5 g | |
1-hydroxycyclohexyl phenyl ketone, 1-hydroxycyclohexyl phenyl methanone, 1-benzoylcyclohexanol, hydroxycyclohexyl phenyl ketone, 1-benzoylcyclohexan-1-ol, irgacure 184, methanone, 1-hydroxycyclohexyl phenyl, unii-e7jvn2243x, 1-hydroxycyclohexyl-phenylmethanone, 1-cyclohexylhydroxyphenylketone | |
QNODIIQQMGDSEF-UHFFFAOYSA-N | |
(1-hydroxycyclohexyl)-phenylmethanone | |
70355 | |
98% |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Warning
EINECSNumber : 213-426-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only