missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(1-Imidazolyl)acetonitrile 98.0+%, TCI America™
$276.51 - $821.60
Chemical Identifiers
CAS | 98873-55-3 |
---|---|
Molecular Formula | C5H5N3 |
Molecular Weight (g/mol) | 107.116 |
MDL Number | MFCD06421433 |
InChI Key | ZPGCVVBPGQJSPX-UHFFFAOYSA-N |
Synonym | 2-1h-imidazol-1-yl acetonitrile, imidazol-1-yl-acetonitrile, 1-cyanomethyl imidazole, 1h-imidazole-1-acetonitrile, 1h-imidazol-1-ylacetonitrile, 2-1-imidazolyl acetonitrile, 1-imidazolyl acetonitrile, 2-imidazol-1-yl acetonitrile, 1h-imidazol-1-yl acetonitrile, 1-cyanomethylimidazole |
PubChem CID | 11804695 |
IUPAC Name | 2-imidazol-1-ylacetonitrile |
SMILES | C1=CN(C=N1)CC#N |
Chemical Identifiers
98873-55-3 | |
107.116 | |
ZPGCVVBPGQJSPX-UHFFFAOYSA-N | |
11804695 | |
C1=CN(C=N1)CC#N |
C5H5N3 | |
MFCD06421433 | |
2-1h-imidazol-1-yl acetonitrile, imidazol-1-yl-acetonitrile, 1-cyanomethyl imidazole, 1h-imidazole-1-acetonitrile, 1h-imidazol-1-ylacetonitrile, 2-1-imidazolyl acetonitrile, 1-imidazolyl acetonitrile, 2-imidazol-1-yl acetonitrile, 1h-imidazol-1-yl acetonitrile, 1-cyanomethylimidazole | |
2-imidazol-1-ylacetonitrile |
Specifications
98873-55-3 | |
C5H5N3 | |
1 g | |
2-1h-imidazol-1-yl acetonitrile, imidazol-1-yl-acetonitrile, 1-cyanomethyl imidazole, 1h-imidazole-1-acetonitrile, 1h-imidazol-1-ylacetonitrile, 2-1-imidazolyl acetonitrile, 1-imidazolyl acetonitrile, 2-imidazol-1-yl acetonitrile, 1h-imidazol-1-yl acetonitrile, 1-cyanomethylimidazole | |
C1=CN(C=N1)CC#N | |
107.116 | |
107.12 | |
Crystalline Powder |
56°C | |
MFCD06421433 | |
3439 | |
ZPGCVVBPGQJSPX-UHFFFAOYSA-N | |
2-imidazol-1-ylacetonitrile | |
11804695 | |
≥98.0% (GC,T) | |
(1-Imidazolyl)acetonitrile |
Safety and Handling
TSCA : No