missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Iodo-3,5-bis(trifluoromethyl)benzene 98.0+%, TCI America™
Supplier: TCI America B186825G
Specifications
1-Iodo-3,5-bis(trifluoromethyl)benzene | |
Yellow | |
C8H3F6I | |
25 g | |
VDPIZIZDKPFXLI-UHFFFAOYSA-N | |
1-iodo-3,5-bis(trifluoromethyl)benzene | |
630970 | |
≥98.0% (GC) |
328-73-4 | |
172°C | |
MFCD00040837 | |
1-iodo-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl iodobenzene, 1,3-bis trifluoromethyl-5-iodobenzene, 3,5-bis trifluoromethyl-1-iodo benzene, benzene, 1-iodo-3,5-bis trifluoromethyl, 1-iodo-3,5-di trifluoromethyl benzene, 3-5-di trifluoromethyl iodobenzene, pubchem1056, acmc-209hwp | |
C1=C(C=C(C=C1C(F)(F)F)I)C(F)(F)F | |
340.007 | |
340.01 | |
Liquid |
Chemical Identifiers
328-73-4 | |
340.007 | |
VDPIZIZDKPFXLI-UHFFFAOYSA-N | |
630970 | |
C1=C(C=C(C=C1C(F)(F)F)I)C(F)(F)F |
C8H3F6I | |
MFCD00040837 | |
1-iodo-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl iodobenzene, 1,3-bis trifluoromethyl-5-iodobenzene, 3,5-bis trifluoromethyl-1-iodo benzene, benzene, 1-iodo-3,5-bis trifluoromethyl, 1-iodo-3,5-di trifluoromethyl benzene, 3-5-di trifluoromethyl iodobenzene, pubchem1056, acmc-209hwp | |
1-iodo-3,5-bis(trifluoromethyl)benzene |
Safety and Handling
TSCA : No