missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Iodobutane, 98%, stabilized, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 173431000

 View more versions of this product

Catalog No. AC173431000

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Iodobutane
542-69-8,7440-50-8
100.0
Colorless to Yellow or Pink
130.0°C to 131.0°C
Authentic
Glass bottle
1.4975 to 1.4995
MFCD00001098
01, 123
15, 1576
Solubility in water: practically insoluble. Other solubilities: soluble in alcohol and ether
CCCCI
184.02
10962
98%
98%
97.5
-103.0°C
1.6100g/mL
33°C
97.5% min. (GC)
C4H9I
CH3(CH2)3I
100 mL
1.61
butyl iodide, butane, 1-iodo, n-butyl iodide, 1-iodo-butane, butane, iodo, iodobutane, 1-jodbutan, 1-jodbutan czech, unii-607a6cc46r, butyliodide
KMGBZBJJOKUPIA-UHFFFAOYSA-N
1-iodobutane
copper
184.02
Liquid

Chemical Identifiers

542-69-8
184.02
KMGBZBJJOKUPIA-UHFFFAOYSA-N
10962
CCCCI
C4H9I
MFCD00001098
butyl iodide, butane, 1-iodo, n-butyl iodide, 1-iodo-butane, butane, iodo, iodobutane, 1-jodbutan, 1-jodbutan czech, unii-607a6cc46r, butyliodide
1-iodobutane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protectio

GHS Signal Word: Danger

EINECSNumber : 208-824-4

RTECSNumber : EK4400000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only