Learn More
1-(Mercaptomethyl)cyclopropaneacetic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6172303
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-(Mercaptomethyl)cyclopropaneacetic acid | |
42°C to 46°C | |
>110°C (230°F) | |
C6H10O2S | |
1 g | |
2-1-mercaptomethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropaneacetic acid, 1-mercaptomethyl cyclopropylacetic acid, 1-mercaptomethyl-cyclopropaneacetic acid, unii-1x00t93wyx, cyclopropaneacetic acid, 1-mercaptomethyl, 2-1-sulfanylmethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropyl acetic acid, 1-sulfanylmethyl cyclopropyl acetic acid | |
C1CC1(CC(=O)O)CS | |
146.204 | |
146.21 | |
Crystalline |
162515-68-6 | |
White | |
Stench | |
MFCD03411600 | |
UN3261 | |
VFAXPOVKNPTBTM-UHFFFAOYSA-N | |
2-[1-(sulfanylmethyl)cyclopropyl]acetic acid | |
9793825 | |
98% |
Chemical Identifiers
162515-68-6 | |
146.204 | |
VFAXPOVKNPTBTM-UHFFFAOYSA-N | |
9793825 | |
C1CC1(CC(=O)O)CS |
C6H10O2S | |
MFCD03411600 | |
2-1-mercaptomethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropaneacetic acid, 1-mercaptomethyl cyclopropylacetic acid, 1-mercaptomethyl-cyclopropaneacetic acid, unii-1x00t93wyx, cyclopropaneacetic acid, 1-mercaptomethyl, 2-1-sulfanylmethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropyl acetic acid, 1-sulfanylmethyl cyclopropyl acetic acid | |
2-[1-(sulfanylmethyl)cyclopropyl]acetic acid |
Safety and Handling
GHS H Statement
H314-H302-H312-H317
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H302+H312-H314-H317-H335
DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 420-240-3
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen