Learn More
1-Methoxy-2-methyl-2-propanol, 98+%, Thermo Scientific Chemicals
$482.11 - $1574.16
Chemical Identifiers
CAS | 3587-64-2 |
---|---|
Molecular Formula | C5H12O2 |
Molecular Weight (g/mol) | 104.15 |
MDL Number | MFCD03701584 |
InChI Key | MXUXZWFVAPTPAG-UHFFFAOYSA-N |
Synonym | 1-methoxy-2-methyl-2-propanol, 2-propanol, 1-methoxy-2-methyl, acmc-20aott, 2-methoxymethyl-2-propanol, 1,1-dimethyl-2-methoxyethanol, 2-propanol,1-methoxy-2-methyl, 1-methoxy-2-methyl-2-propanol # |
PubChem CID | 77137 |
IUPAC Name | 1-methoxy-2-methylpropan-2-ol |
SMILES | COCC(C)(C)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL2035114
|
Thermo Scientific Chemicals
L2035114 |
25 g |
Each for $482.11
|
|
||||||
AAL2035122
|
Thermo Scientific Chemicals
L2035122 |
100 g |
Each for $1,574.16
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3587-64-2 | |
104.15 | |
MXUXZWFVAPTPAG-UHFFFAOYSA-N | |
77137 | |
COCC(C)(C)O |
C5H12O2 | |
MFCD03701584 | |
1-methoxy-2-methyl-2-propanol, 2-propanol, 1-methoxy-2-methyl, acmc-20aott, 2-methoxymethyl-2-propanol, 1,1-dimethyl-2-methoxyethanol, 2-propanol,1-methoxy-2-methyl, 1-methoxy-2-methyl-2-propanol # | |
1-methoxy-2-methylpropan-2-ol |
Specifications
3587-64-2 | |
115°C to 116°C | |
C5H12O2 | |
MFCD03701584 | |
UN1987 | |
MXUXZWFVAPTPAG-UHFFFAOYSA-N | |
1-methoxy-2-methylpropan-2-ol | |
77137 | |
≥98% |
0.892 | |
27°C (80°F) | |
1.4044 | |
25 g | |
1-methoxy-2-methyl-2-propanol, 2-propanol, 1-methoxy-2-methyl, acmc-20aott, 2-methoxymethyl-2-propanol, 1,1-dimethyl-2-methoxyethanol, 2-propanol,1-methoxy-2-methyl, 1-methoxy-2-methyl-2-propanol # | |
COCC(C)(C)O | |
104.15 | |
104.15 | |
1-Methoxy-2-methyl-2-propanol |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only