missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methoxy-2-propanol 98.0+%, TCI America™

Supplier:  TCI America M012625ML

 View more versions of this product

Catalog No. M012625ML


Explore available promotions

Description

Description

Specifications

Specifications

1-Methoxy-2-propanol
-96°C
120°C
MFCD00004537
3092
ARXJGSRGQADJSQ-UHFFFAOYNA-N
1-methoxypropan-2-ol
7900
≥98.0% (GC)
107-98-2
Colorless
C4H10O2
25 mL
1-methoxy-2-propanol, pgme, methoxyisopropanol, 2-propanol, 1-methoxy, closol, dowtherm 209, propylene glycol monomethyl ether, poly-solve mpm, propasol solvent m, dowanol 33b
COCC(C)O
90.12
90.12
Liquid

Chemical Identifiers

107-98-2
90.12
ARXJGSRGQADJSQ-UHFFFAOYNA-N
7900
COCC(C)O
C4H10O2
MFCD00004537
1-methoxy-2-propanol, pgme, methoxyisopropanol, 2-propanol, 1-methoxy, closol, dowtherm 209, propylene glycol monomethyl ether, poly-solve mpm, propasol solvent m, dowanol 33b
1-methoxypropan-2-ol
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0404

RTECSNumber : UB7700000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions