missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-4-methylcyclohexane (cis- and trans- mixture) 98.0+%, TCI America™

$271.78 - $921.03

Chemical Identifiers

CAS 90200-72-9
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
InChI Key VBAPYXHNMYKBKN-UHFFFAOYSA-N
PubChem CID 6336264
IUPAC Name 1-methoxy-4-methylcyclohexane
SMILES CC1CCC(CC1)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M23755G
View Documents
TCI America
M23755G
5 g
Each for $271.78
Only null left
 
M237525G
View Documents
TCI America
M237525G
25 g
Each for $921.03
Only null left
 
Description

Description

Specifications

Chemical Identifiers

90200-72-9
128.215
6336264
CC1CCC(CC1)OC
C8H16O
VBAPYXHNMYKBKN-UHFFFAOYSA-N
1-methoxy-4-methylcyclohexane

Specifications

90200-72-9
148°C
5 g
VBAPYXHNMYKBKN-UHFFFAOYSA-N
1-methoxy-4-methylcyclohexane
6336264
≥98.0% (GC)
1-Methoxy-4-methylcyclohexane (cis- and trans- mixture)
Colorless
C8H16O
3271
CC1CCC(CC1)OC
128.215
128.22
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'