missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Methyl-1,2-epoxycyclohexane 95.0+%, TCI America™
$246.87 - $697.17
Chemical Identifiers
CAS | 1713-33-3 |
---|---|
Molecular Formula | C7H12O |
Molecular Weight (g/mol) | 112.17 |
MDL Number | MFCD11846121 |
InChI Key | FIEKVYPYFQSFTP-UHFFFAOYNA-N |
Synonym | 1,2-Epoxy-1-methylcyclohexane, 1-Methyl-7-oxabicyclo[4.1.0]heptane |
PubChem CID | 224243 |
IUPAC Name | 1-methyl-7-oxabicyclo[4.1.0]heptane |
SMILES | CC12CCCCC1O2 |
Chemical Identifiers
1713-33-3 | |
112.17 | |
FIEKVYPYFQSFTP-UHFFFAOYNA-N | |
224243 | |
CC12CCCCC1O2 |
C7H12O | |
MFCD11846121 | |
1,2-Epoxy-1-methylcyclohexane, 1-Methyl-7-oxabicyclo[4.1.0]heptane | |
1-methyl-7-oxabicyclo[4.1.0]heptane |
Specifications
1713-33-3 | |
138°C | |
MFCD11846121 | |
1993 | |
FIEKVYPYFQSFTP-UHFFFAOYNA-N | |
1-methyl-7-oxabicyclo[4.1.0]heptane | |
224243 | |
≥95.0% (GC) | |
1-Methyl-1,2-epoxycyclohexane |
Yellow | |
C7H12O | |
1 g | |
1,2-Epoxy-1-methylcyclohexane, 1-Methyl-7-oxabicyclo[4.1.0]heptane | |
CC12CCCCC1O2 | |
112.17 | |
112.17 | |
Liquid |
Safety and Handling
TSCA : No