missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-1H-imidazole-4-carbaldehyde, 95%, Thermo Scientific™
$425.85 - $425.85
Chemical Identifiers
| CAS | 17289-26-8 |
|---|---|
| Molecular Formula | C5H6N2O |
| Molecular Weight (g/mol) | 110.116 |
| MDL Number | MFCD03411957 |
| InChI Key | CQZXDIHVSPZIGF-UHFFFAOYSA-N |
| PubChem CID | 573419 |
| IUPAC Name | 1-methylimidazole-4-carbaldehyde |
| SMILES | CN1C=C(N=C1)C=O |
Chemical Identifiers
| 17289-26-8 | |
| 110.116 | |
| CQZXDIHVSPZIGF-UHFFFAOYSA-N | |
| 1-methylimidazole-4-carbaldehyde |
| C5H6N2O | |
| MFCD03411957 | |
| 573419 | |
| CN1C=C(N=C1)C=O |
Specifications
| 17289-26-8 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD03411957 | |
| 1-methyl-1h-imidazole-4-carbaldehyde, 1-methylimidazole-4-carboxaldehyde, 1-methyl-1h-imidazole-4-carboxaldehyde, 1h-imidazole-4-carboxaldehyde, 1-methyl, methyl-1h-imidazole-4-carbaldehyde, 1-methylimidazol-4-carboxaldehyde, 4-formyl-1-methyl-1h-imidazole, 1-methyl-4-formyl-imidazole, 1-methyl-1h-imidazole-4-carbaldehyde #, 1h-imidazole-4-carboxaldehyde,1-methyl | |
| CN1C=C(N=C1)C=O | |
| 110.116 | |
| 110.12 | |
| 1-Methyl-1H-imidazole-4-carbaldehyde |
| 94.5 | |
| 95% | |
| C5H6N2O | |
| 250 mg | |
| CQZXDIHVSPZIGF-UHFFFAOYSA-N | |
| 1-methylimidazole-4-carbaldehyde | |
| 573419 | |
| 95% |
Safety and Handling
GHS P Statement Causes skin irritation. Harmful if swallowed. May cause respiratory irritation. Causes serious eye irritation.
GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protec
Warning