missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-1H-imidazole-4-sulfonamide, 97%, Thermo Scientific™

Chemical Identifiers

CAS 111124-90-4
Molecular Formula C4H7N3O2S
Molecular Weight (g/mol) 161.18
MDL Number MFCD02091382
InChI Key QBJSSOBNVYDCDQ-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazole-4-sulfonamide, 1h-imidazole-4-sulfonamide,1-methyl, 1-methyl-1h-imidazole-4-sulphonamide, acmc-20dtvv, maybridge1_000050, spr_7, 1-methyl-4-imidazolesulfonamide, 1-methyl-1h-imidazol-4-ylsulfonamide, 1h-imidazole-4-sulfonamide, 1-methyl
PubChem CID 2736885
IUPAC Name 1-methylimidazole-4-sulfonamide
SMILES CN1C=NC(=C1)S(N)(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
TL01003CB
SDS View Documents
Thermo Scientific Chemicals
TL01003CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

111124-90-4
161.18
QBJSSOBNVYDCDQ-UHFFFAOYSA-N
2736885
CN1C=NC(=C1)S(N)(=O)=O
C4H7N3O2S
MFCD02091382
1-methyl-1h-imidazole-4-sulfonamide, 1h-imidazole-4-sulfonamide,1-methyl, 1-methyl-1h-imidazole-4-sulphonamide, acmc-20dtvv, maybridge1_000050, spr_7, 1-methyl-4-imidazolesulfonamide, 1-methyl-1h-imidazol-4-ylsulfonamide, 1h-imidazole-4-sulfonamide, 1-methyl
1-methylimidazole-4-sulfonamide

Specifications

111124-90-4
97%
C4H7N3O2S
250mg
QBJSSOBNVYDCDQ-UHFFFAOYSA-N
1-methylimidazole-4-sulfonamide
2736885
97%
97.0
Amber Glass Bottle
MFCD02091382
1-methyl-1h-imidazole-4-sulfonamide, 1h-imidazole-4-sulfonamide,1-methyl, 1-methyl-1h-imidazole-4-sulphonamide, acmc-20dtvv, maybridge1_000050, spr_7, 1-methyl-4-imidazolesulfonamide, 1-methyl-1h-imidazol-4-ylsulfonamide, 1h-imidazole-4-sulfonamide, 1-methyl
CN1C=NC(=C1)S(N)(=O)=O
161.18
161.18
1-Methyl-1H-imidazole-4-sulfonamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
missing translation for 'documents'

missing translation for 'documents'