missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indole-5-carbaldehyde, 97%, Thermo Scientific™

$405.14 - $405.14

Chemical Identifiers

CAS 90923-75-4
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD08271908
InChI Key XIVDZBIBWGQOTI-UHFFFAOYSA-N
PubChem CID 7537534
SMILES CN1C=CC2=CC(C=O)=CC=C12
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC41404DA
SDS View Documents
Thermo Scientific Chemicals
CC41404DA
1 g
Each for $405.14
Only null left
 
Description

Description

Specifications

Chemical Identifiers

90923-75-4
159.19
XIVDZBIBWGQOTI-UHFFFAOYSA-N
CN1C=CC2=CC(C=O)=CC=C12
C10H9NO
MFCD08271908
7537534

Specifications

90923-75-4
100.0
Amber Glass Bottle
MFCD08271908
1-methyl-1h-indole-5-carbaldehyde, 1h-indole-5-carboxaldehyde, 1-methyl, 5-formyl-1-methylindole, n-methylindol-5-carbaldehyde, n-methylindole-5-carbaldehyde, n-methylindole-5-carboxaldehyde, 1-methyl-1h-indole-5-carboxaldehyde, 1h-indole-5-carboxaldehyde,1-methyl
CN1C=CC2=CC(C=O)=CC=C12
159.19
159.19
1-Methyl-1H-indole-5-carbaldehyde
96.5
97%
C10H9NO
1 g
XIVDZBIBWGQOTI-UHFFFAOYSA-N
1-methylindole-5-carbaldehyde
7537534
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'