missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Methyl 2-Aminoterephthalate 98.0+%, TCI America™
Supplier: TCI America A15075G
Specifications
1-Methyl 2-Aminoterephthalate | |
218°C | |
C9H9NO4 | |
5 g | |
QKOKLMFCKLEFDV-UHFFFAOYSA-N | |
3-amino-4-methoxycarbonylbenzoic acid | |
5235500 | |
≥98.0% (T) |
60728-41-8 | |
Yellow | |
MFCD00189374 | |
3-Amino-4-carbomethoxybenzoic Acid, 3-Amino-4-methoxycarbonylbenzoic Acid, 2-Aminoterephthalic Acid 1-Methyl Ester | |
COC(=O)C1=C(C=C(C=C1)C(=O)O)N | |
195.174 | |
195.17 | |
Crystalline Powder |
Chemical Identifiers
60728-41-8 | |
195.174 | |
QKOKLMFCKLEFDV-UHFFFAOYSA-N | |
5235500 | |
COC(=O)C1=C(C=C(C=C1)C(=O)O)N |
C9H9NO4 | |
MFCD00189374 | |
3-Amino-4-carbomethoxybenzoic Acid, 3-Amino-4-methoxycarbonylbenzoic Acid, 2-Aminoterephthalic Acid 1-Methyl Ester | |
3-amino-4-methoxycarbonylbenzoic acid |
Safety and Handling
TSCA : No