missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyridone 99.0+%, TCI America™

$242.33 - $417.56

Chemical Identifiers

CAS 694-85-9
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00006520
InChI Key DVVGIUUJYPYENY-UHFFFAOYSA-N
Synonym 1-methyl-2-pyridone, 1-methylpyridin-2 1h-one, n-methyl-2-pyridone, 2 1h-pyridinone, 1-methyl, 1-methyl-2 1h-pyridinone, n-methylpyridone, 1-methyl-2 1h-pyridone, 1-methyl-2-pyridinone, 2 1h-pyridone, 1-methyl, 1-methyl-1,2-dihydro-2-pyridinone
PubChem CID 12755
IUPAC Name 1-methylpyridin-2-one
SMILES CN1C=CC=CC1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M041210G
View Documents
TCI America
M041210G
10 g
Each for $242.33
Only null left
 
M041225G
View Documents
TCI America
M041225G
25 g
Each for $417.56
Only null left
 
Description

Description

Specifications

Chemical Identifiers

694-85-9
109.128
DVVGIUUJYPYENY-UHFFFAOYSA-N
12755
CN1C=CC=CC1=O
C6H7NO
MFCD00006520
1-methyl-2-pyridone, 1-methylpyridin-2 1h-one, n-methyl-2-pyridone, 2 1h-pyridinone, 1-methyl, 1-methyl-2 1h-pyridinone, n-methylpyridone, 1-methyl-2 1h-pyridone, 1-methyl-2-pyridinone, 2 1h-pyridone, 1-methyl, 1-methyl-1,2-dihydro-2-pyridinone
1-methylpyridin-2-one

Specifications

694-85-9
252°C
MFCD00006520
1-methyl-2-pyridone, 1-methylpyridin-2 1h-one, n-methyl-2-pyridone, 2 1h-pyridinone, 1-methyl, 1-methyl-2 1h-pyridinone, n-methylpyridone, 1-methyl-2 1h-pyridone, 1-methyl-2-pyridinone, 2 1h-pyridone, 1-methyl, 1-methyl-1,2-dihydro-2-pyridinone
CN1C=CC=CC1=O
109.128
109.13
Liquid
Yellow
C6H7NO
10 g
DVVGIUUJYPYENY-UHFFFAOYSA-N
1-methylpyridin-2-one
12755
≥99.0% (GC)
1-Methyl-2-pyridone
Videos
Safety and Handling

Safety and Handling

RTECSNumber : UV1148100

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'