missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidinone, 98%, pure

1-Methyl-2-pyrrolidinone, 98%, C5H9NO, CAS Number-872-50-4, 25L, 97.5% min. (GC), 202.0 deg.C, 1.0300g/mL, 212-828-1, 91 deg.C, 99.13, Pure, GHS Signal Word: Danger, SECXISVLQFMRJM-UHFFFAOYSA-N, 1-methylpyrrolidin-2-one, -24.0 deg.C, 15,619, Metal drum, 1.4690 to 1.4710, CN1CCCC1-O | CAS: 872-50-4 | C5H9NO | 99.13 g/mol

Supplier:  Thermo Scientific Chemicals 449180250

 View more versions of this product

Catalog No. AC449180250

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Specifications

1-Methyl-2-pyrrolidinone
872-50-4
202.0°C
91°C
1.4690 to 1.4710
1.03
1.67 mPa.s (20°C)
Solubility in water: 1000g/L (20°C). Other solubilities: miscible with most common organic solvents
CN1CCCC1=O
99.13
98%
-24.0°C
1.0300g/mL
25 L
Metal Drum
97.5% min. (GC)
C5H9NO
15,619
SECXISVLQFMRJM-UHFFFAOYSA-N
1-methylpyrrolidin-2-one
99.13
Pure

Chemical Identifiers

872-50-4
99.13
1-methylpyrrolidin-2-one
C5H9NO
SECXISVLQFMRJM-UHFFFAOYSA-N
CN1CCCC1=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 212-828-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only