missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals

$99.63 - $1105.92

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
Synonym 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
PubChem CID 13387
ChEBI CHEBI:7307
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC390682500
SDS View Documents
Thermo Scientific Chemicals
390682500
250 mL Glass Bottle
Each for $99.63
 
AC390680010
SDS View Documents
Thermo Scientific Chemicals
390680010
1 L Glass Bottle
Each for $197.25
 
AC390680025
SDS View Documents
Thermo Scientific Chemicals
390680025
2.5 L Glass Bottle
Each for $519.93
 
AC390680100
SDS View Documents
Thermo Scientific Chemicals
390680100
10 L Plastic Drum
Each for $1,105.92
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Identifiers

872-50-4
99.133
1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
CHEBI:7307
CN1CCCC1=O
C5H9NO
SECXISVLQFMRJM-UHFFFAOYSA-N
13387
1-methylpyrrolidin-2-one

Specifications

-24.0°C
1.0300g/mL
250 mL
Authentic
Liquid
1.03
1.67 mPa.s (20°C)
01,696; 02,281; 09,316; 11,346
Solubility in water: 1000g/L (20°C). Other solubilities: miscible with most common organic solvents
CN1CCCC1=O
99.133
CHEBI:7307
≥99.5%
1-Methyl-2-pyrrolidinone
872-50-4
202.0°C
91°C
Glass Bottle
1.4690 to 1.4710
C5H9NO
0.05% max.
1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
SECXISVLQFMRJM-UHFFFAOYSA-N
1-methylpyrrolidin-2-one
13387
99.13
ACS Reagent
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
May damage the unborn child.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only