missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-2-(tri-n-butylstannyl)imidazole, 90+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5152303

 View more versions of this product

Catalog No. AAH5152303


Explore available promotions

Description

Description

1-Methyl-2-(tri-n-butylstannyl)imidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials. It is an organostannic compound for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methyl-2-(tri-n-butylstannyl)imidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials. It is an organostannic compound for proteomics research.

Solubility
Sparingly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications

Specifications

1-Methyl-2-(tri-n-butylstannyl)imidazole
C16H32N2Sn
1 g
1-methyl-2-tributylstannyl-1h-imidazole, 1-methyl-2-tributylstannyl imidazole, 1-methyl-2-tri-n-butylstannyl imidazole, 1-methyl-1h-imidazol-2-yl tributylstannane, 1-methyl-2-tributylstannanyl-1h-imidazole, acmc-20aotl, tributyl-1-methylimidazol-2-yl stannane, n-methyl 2-tributylstannylimidazole, 2-tri-n-butylstannyl-n-methylimidazole
KFWFYOPKNSYTMV-UHFFFAOYSA-N
tributyl-(1-methylimidazol-2-yl)stannane
2763252
≥90%
105494-69-7
MFCD01319030
UN2788
Sparingly soluble in water.
CCCC[Sn](CCCC)(CCCC)C1=NC=CN1C
371.156
371.15

Chemical Identifiers

105494-69-7
371.156
KFWFYOPKNSYTMV-UHFFFAOYSA-N
2763252
CCCC[Sn](CCCC)(CCCC)C1=NC=CN1C
C16H32N2Sn
MFCD01319030
1-methyl-2-tributylstannyl-1h-imidazole, 1-methyl-2-tributylstannyl imidazole, 1-methyl-2-tri-n-butylstannyl imidazole, 1-methyl-1h-imidazol-2-yl tributylstannane, 1-methyl-2-tributylstannanyl-1h-imidazole, acmc-20aotl, tributyl-1-methylimidazol-2-yl stannane, n-methyl 2-tributylstannylimidazole, 2-tri-n-butylstannyl-n-methylimidazole
tributyl-(1-methylimidazol-2-yl)stannane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c

H301-H312-H315-H319-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only