Learn More
1-Methylcyclohexanecarboxylic acid, 99%, Thermo Scientific Chemicals
$86.58 - $498.55
Chemical Identifiers
CAS | 1123-25-7 |
---|---|
Molecular Formula | C8H14O2 |
Molecular Weight (g/mol) | 142.198 |
MDL Number | MFCD00001463 |
InChI Key | REHQLKUNRPCYEW-UHFFFAOYSA-N |
Synonym | 1-methylcyclohexanecarboxylic acid, 1-methyl-1-cyclohexanecarboxylic acid, 1-methylcyclohexylcarboxylic acid, 1-methylcyclohexanoic acid, cyclohexanecarboxylic acid, 1-methyl, 1-methyl-1-cyclohexanoic acid, 1-methyl-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, methyl, acmc-2099ec, 4-09-00-00041 beilstein handbook reference |
PubChem CID | 70744 |
IUPAC Name | 1-methylcyclohexane-1-carboxylic acid |
SMILES | CC1(CCCCC1)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAB2171306
|
Thermo Scientific Chemicals
B2171306 |
5 g |
N/A
|
|
||||||
AAB2171314
|
Thermo Scientific Chemicals
B2171314 |
25 g |
N/A
|
|
||||||
AAB2171318
|
Thermo Scientific Chemicals
B2171318 |
50 g |
N/A
|
|
||||||
Description
1-Methyl-1-cyclohexanecarboxylic acid was used as internal standard during the determination of valproic acid metabolites. Also been used as an anticonvulsant drug and causes maturation of murine neuroblastoma cells in vitro. Used in the synthesis of p38 map kinase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in chloroform and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
1123-25-7 | |
142.198 | |
REHQLKUNRPCYEW-UHFFFAOYSA-N | |
70744 | |
CC1(CCCCC1)C(=O)O |
C8H14O2 | |
MFCD00001463 | |
1-methylcyclohexanecarboxylic acid, 1-methyl-1-cyclohexanecarboxylic acid, 1-methylcyclohexylcarboxylic acid, 1-methylcyclohexanoic acid, cyclohexanecarboxylic acid, 1-methyl, 1-methyl-1-cyclohexanoic acid, 1-methyl-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, methyl, acmc-2099ec, 4-09-00-00041 beilstein handbook reference | |
1-methylcyclohexane-1-carboxylic acid |
Specifications
1123-25-7 | |
236°C | |
C8H14O2 | |
5 g | |
1-methylcyclohexanecarboxylic acid, 1-methyl-1-cyclohexanecarboxylic acid, 1-methylcyclohexylcarboxylic acid, 1-methylcyclohexanoic acid, cyclohexanecarboxylic acid, 1-methyl, 1-methyl-1-cyclohexanoic acid, 1-methyl-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, methyl, acmc-2099ec, 4-09-00-00041 beilstein handbook reference | |
REHQLKUNRPCYEW-UHFFFAOYSA-N | |
1-methylcyclohexane-1-carboxylic acid | |
70744 | |
99% |
36°C to 39°C | |
100°C (212°F) | |
MFCD00001463 | |
1859220 | |
Soluble in chloroform and methanol. | |
CC1(CCCCC1)C(=O)O | |
142.198 | |
142.2 | |
1-Methylcyclohexanecarboxylic acid |
Safety and Handling
EINECSNumber : 214-371-3
RTECSNumber : GU8585500
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only