Learn More
1-Methylindole-2-boronic acid, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5302803
Description
Reactant for Suzuki-Miyaura coupling. It ia an active Pharmaceutical ingredient hence used as pharmaceutical intermediates. Also used as rawv material in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReactant for Suzuki-Miyaura coupling. It ia an active Pharmaceutical ingredient hence used as pharmaceutical intermediates. Also used as rawv material in organic synthesis.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
Specifications
1-Methylindole-2-boronic acid | |
108°C to 112°C | |
MFCD01114668 | |
1-methyl-1h-indol-2-yl boronic acid, n-methylindole-2-boronic acid, 2-borono-1-methyl-1h-indole, 1-methylindol-2-yl boronic acid, 1-methyl-1h-indole-2-boronic acid, 1-methyl-1h-indol-2-yl-2-boronic acid, boronic acid,b-1-methyl-1h-indol-2-yl, boronic acid, 1-methyl-1h-indol-2-yl, acmc-209eur, n-methyl-indoleboronic acid | |
CN1C(=CC2=CC=CC=C12)B(O)O | |
174.99 | |
174.99 |
191162-40-0 | |
C9H10BNO2 | |
1 g | |
CBPBJUTWVXLSER-UHFFFAOYSA-N | |
(1-methylindol-2-yl)boronic acid | |
22733820 | |
95% |
Chemical Identifiers
191162-40-0 | |
174.99 | |
CBPBJUTWVXLSER-UHFFFAOYSA-N | |
22733820 |
C9H10BNO2 | |
MFCD01114668 | |
1-methyl-1h-indol-2-yl boronic acid, n-methylindole-2-boronic acid, 2-borono-1-methyl-1h-indole, 1-methylindol-2-yl boronic acid, 1-methyl-1h-indole-2-boronic acid, 1-methyl-1h-indol-2-yl-2-boronic acid, boronic acid,b-1-methyl-1h-indol-2-yl, boronic acid, 1-methyl-1h-indol-2-yl, acmc-209eur, n-methyl-indoleboronic acid | |
CN1C(=CC2=CC=CC=C12)B(O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only