Learn More
1-Methylpyrrole-2-carboxylic acid, 98%, Thermo Scientific Chemicals
$62.58 - $198.56
Chemical Identifiers
CAS | 6973-60-0 |
---|---|
Molecular Formula | C6H7NO2 |
Molecular Weight (g/mol) | 125.127 |
MDL Number | MFCD00003088 |
InChI Key | ILAOVOOZLVGAJF-UHFFFAOYSA-N |
Synonym | 1-methyl-1h-pyrrole-2-carboxylic acid, 1-methyl-2-pyrrolecarboxylic acid, n-methylpyrrole-2-carboxylic acid, 1h-pyrrole-2-carboxylic acid, 1-methyl, 1h-pyrrolecarboxylic acid, methyl, 1-methyl-1h-pyrrole-2-carboxylicacid, pubchem9181, acmc-209oar, ksc353i0t, 2-carboxy-1-methyl-1h-pyrrole |
PubChem CID | 81453 |
IUPAC Name | 1-methylpyrrole-2-carboxylic acid |
SMILES | CN1C=CC=C1C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0590603
|
Thermo Scientific Chemicals
L0590603 |
1 g |
Each for $62.58
|
|
|||||
AAL0590606
|
Thermo Scientific Chemicals
L0590606 |
5 g |
Each for $198.56
|
|
|||||
Description
1-Methylpyrrole-2-carboxylic acid is used for thermochemical and computational studies of 1-Methyl-2-pyrrolecarboxylic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Methylpyrrole-2-carboxylic acid is used for thermochemical and computational studies of 1-Methyl-2-pyrrolecarboxylic acid.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
Chemical Identifiers
6973-60-0 | |
125.127 | |
ILAOVOOZLVGAJF-UHFFFAOYSA-N | |
81453 | |
CN1C=CC=C1C(=O)O |
C6H7NO2 | |
MFCD00003088 | |
1-methyl-1h-pyrrole-2-carboxylic acid, 1-methyl-2-pyrrolecarboxylic acid, n-methylpyrrole-2-carboxylic acid, 1h-pyrrole-2-carboxylic acid, 1-methyl, 1h-pyrrolecarboxylic acid, methyl, 1-methyl-1h-pyrrole-2-carboxylicacid, pubchem9181, acmc-209oar, ksc353i0t, 2-carboxy-1-methyl-1h-pyrrole | |
1-methylpyrrole-2-carboxylic acid |
Specifications
6973-60-0 | |
C6H7NO2 | |
1 g | |
1-methyl-1h-pyrrole-2-carboxylic acid, 1-methyl-2-pyrrolecarboxylic acid, n-methylpyrrole-2-carboxylic acid, 1h-pyrrole-2-carboxylic acid, 1-methyl, 1h-pyrrolecarboxylic acid, methyl, 1-methyl-1h-pyrrole-2-carboxylicacid, pubchem9181, acmc-209oar, ksc353i0t, 2-carboxy-1-methyl-1h-pyrrole | |
CN1C=CC=C1C(=O)O | |
125.127 | |
125.13 | |
1-Methylpyrrole-2-carboxylic acid |
135°C to 138°C | |
MFCD00003088 | |
115703 | |
ILAOVOOZLVGAJF-UHFFFAOYSA-N | |
1-methylpyrrole-2-carboxylic acid | |
81453 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 230-217-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only