missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methylurea 98.0+%, TCI America™

Supplier:  TCI America M0455500G

 View more versions of this product

Catalog No. M0455500G


Explore available promotions

Description

Description

Specifications

Specifications

1-Methylurea
99°C
C2H6N2O
500 g
XGEGHDBEHXKFPX-UHFFFAOYSA-N
methylurea
11719
74.08
Crystalline Powder
598-50-5
White
MFCD00007950
1-methylurea, n-methylurea, monomethylurea, urea, methyl, methyl urea, urea, n-methyl, methylmocovina, methylmocovina czech, methylharnstoff german, n-methyl urea
CNC(=O)N
74.083
CHEBI:44383
≥98.0% (N)

Chemical Identifiers

598-50-5
74.083
XGEGHDBEHXKFPX-UHFFFAOYSA-N
11719
methylurea
C2H6N2O
MFCD00007950
1-methylurea, n-methylurea, monomethylurea, urea, methyl, methyl urea, urea, n-methyl, methylmocovina, methylmocovina czech, methylharnstoff german, n-methyl urea
CHEBI:44383
CNC(=O)N
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3254

RTECSNumber : YT7175000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions