missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Naphthyl butyrate, 98%, Thermo Scientific Chemicals
$206.11 - $206.11
Chemical Identifiers
CAS | 3121-70-8 |
---|---|
Molecular Formula | C14H14O2 |
Molecular Weight (g/mol) | 214.26 |
MDL Number | MFCD00003923 |
InChI Key | XIVJNRXPRQKFRZ-UHFFFAOYSA-N |
Synonym | 1-naphthyl butyrate, .alpha.-naphthyl butyrate, alpha-naphthyl butyrate, naphthalen-1-yl butyrate, butanoic acid, 1-naphthalenyl ester, butyric acid, 1-naphthyl ester, 1-naphthylbutyrate, unii-9uzl9u0cik, butyric acid 1-naphthyl ester, 9uzl9u0cik |
PubChem CID | 76571 |
IUPAC Name | naphthalen-1-yl butanoate |
SMILES | CCCC(=O)OC1=CC=CC2=CC=CC=C21 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC334950050
|
Thermo Scientific Chemicals
334950050 |
5 g | Glass Bottle |
Each for $206.11
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3121-70-8 | |
214.26 | |
XIVJNRXPRQKFRZ-UHFFFAOYSA-N | |
76571 | |
CCCC(=O)OC1=CC=CC2=CC=CC=C21 |
C14H14O2 | |
MFCD00003923 | |
1-naphthyl butyrate, .alpha.-naphthyl butyrate, alpha-naphthyl butyrate, naphthalen-1-yl butyrate, butanoic acid, 1-naphthalenyl ester, butyric acid, 1-naphthyl ester, 1-naphthylbutyrate, unii-9uzl9u0cik, butyric acid 1-naphthyl ester, 9uzl9u0cik | |
naphthalen-1-yl butanoate |
Specifications
3121-70-8 | |
100.0 | |
1.0900g/mL | |
62°C | |
97.5% min. (GC) | |
C14H14O2 | |
5 g | |
1-naphthyl butyrate, .alpha.-naphthyl butyrate, alpha-naphthyl butyrate, naphthalen-1-yl butyrate, butanoic acid, 1-naphthalenyl ester, butyric acid, 1-naphthyl ester, 1-naphthylbutyrate, unii-9uzl9u0cik, butyric acid 1-naphthyl ester, 9uzl9u0cik | |
XIVJNRXPRQKFRZ-UHFFFAOYSA-N | |
naphthalen-1-yl butanoate | |
76571 | |
98% | |
1-Naphthyl butyrate, 98% |
97.5 | |
Yellow | |
125°C to 130°C (1.0 mmHg) | |
Authentic | |
Glass Bottle | |
MFCD00003923 | |
1.09 | |
Solubility in water: immiscible in water | |
CCCC(=O)OC1=CC=CC2=CC=CC=C21 | |
214.26 | |
214.26 | |
Liquid |
Safety and Handling
EINECSNumber : 221-500-7
TSCA : TSCA
RUO – Research Use Only