missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Naphthyl isocyanate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 415410250

 View more versions of this product

Catalog No. AC415410250

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Naphthyl isocyanate
98.5
4°C
269°C to 270°C
99%
C11H7NO
25 g
15, 6492
Solubility in water: decomposes. Other solubilities: soluble in toluene,chloroform,ether,decomposes in alcohols
C1=CC=C2C(=C1)C=CC=C2N=C=O
169.18
169.18
86-84-0
100.0
1.1800g/mL
135°C
Glass bottle
1.6324 to 1.6344
1.18
1-naphthyl isocyanate, naphthalene, 1-isocyanato, alpha-naphthyl isocyanate, isocyanatonaphthalene, 1-naphthylisocyanate, 1-isocyanato-naphthalene, unii-5lh2p0691e, 1-isocyanatonapthalene, isocyanic acid 1-naphthyl ester, isocyanic acid, 1-naphthyl ester
BDQNKCYCTYYMAA-UHFFFAOYSA-N
1-isocyanatonaphthalene
66589
99%

Chemical Identifiers

86-84-0
1-naphthyl isocyanate, naphthalene, 1-isocyanato, alpha-naphthyl isocyanate, isocyanatonaphthalene, 1-naphthylisocyanate, 1-isocyanato-naphthalene, unii-5lh2p0691e, 1-isocyanatonapthalene, isocyanic acid 1-naphthyl ester, isocyanic acid, 1-naphthyl ester
1-isocyanatonaphthalene
BDQNKCYCTYYMAA-UHFFFAOYSA-N
66589
C1=CC=C2C(=C1)C=CC=C2N=C=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-703-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only