Learn More
1-Naphthyl phosphate monosodium salt monohydrate, 97+%, Thermo Scientific Chemicals
Substrate for determination of phosphatases
Supplier: Thermo Scientific Chemicals J6300414
Description
Substrate for determination of phosphatases
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Naphthyl phosphate monosodium salt monohydrate | |
White | |
MFCD00150615 | |
1877764 | |
Soluble in water | |
[O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1 | |
222.14 | |
264.15 (246.13 Anhydrous) | |
Powder |
81012-89-7 | |
C10H7O4P | |
25 g | |
1-naphthyl phosphate monosodium salt monohydrate, sodium naphthalen-1-yl hydrogenphosphate hydrate, alpha-naphthyl acid phosphate monosodium, sodium hydrate naphthalen-1-yl hydrogen phosphate, 1-naphthyl phosphate sodium salt monohydrate, c10h8po4.na.h2o, sodium 1-naphthyl phosphate monohydrate, 1-naphthyl phosphate, monosodium salt monohydrate, naphthalen-1-yloxy sodiooxy phosphinic acid hydrate, 1-naphthyl phosphate monosodium salt monohydrate hplc | |
YNXICDMQCQPQEW-UHFFFAOYSA-L | |
naphthalen-1-yl dihydrogen phosphate;sodium;hydrate | |
45055387 | |
≥97% |
Chemical Identifiers
81012-89-7 | |
222.14 | |
YNXICDMQCQPQEW-UHFFFAOYSA-L | |
45055387 |
C10H7O4P | |
MFCD00150615 | |
1-naphthyl phosphate monosodium salt monohydrate, sodium naphthalen-1-yl hydrogenphosphate hydrate, alpha-naphthyl acid phosphate monosodium, sodium hydrate naphthalen-1-yl hydrogen phosphate, 1-naphthyl phosphate sodium salt monohydrate, c10h8po4.na.h2o, sodium 1-naphthyl phosphate monohydrate, 1-naphthyl phosphate, monosodium salt monohydrate, naphthalen-1-yloxy sodiooxy phosphinic acid hydrate, 1-naphthyl phosphate monosodium salt monohydrate hplc | |
[O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only