missing translation for 'onlineSavingsMsg'
Learn More

1-Nitro-2,5-bis(trifluoromethyl)benzene 98.0+%, TCI America™

$48.46 - $145.40

Chemical Identifiers

CAS 320-88-7
Molecular Formula C8H3F6NO2
Molecular Weight (g/mol) 259.107
MDL Number MFCD00236674
InChI Key BZKVUOHHNCMTLH-UHFFFAOYSA-N
Synonym 2,5-bis trifluoromethyl nitrobenzene, 2-nitro-1,4-bis trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-nitrobenzene, 1-nitro-2,5-bis trifluoromethyl benzene, benzene, 2-nitro-1,4-bis trifluoromethyl, 2,5-bistrifluoromethylnitrobenzene, pubchem19738, 2-nitro-1,4-bis-trifluoromethyl-benzene, acmc-1bn6y, ksc496c8f
PubChem CID 2734048
IUPAC Name 2-nitro-1,4-bis(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B25465G
View Documents
TCI America
B25465G
5 g
Each for $48.46
Only null left
 
B254625G
View Documents
TCI America
B254625G
25 g
Each for $145.40
Only null left
 
Description

Description

Specifications

Chemical Identifiers

320-88-7
259.107
BZKVUOHHNCMTLH-UHFFFAOYSA-N
2734048
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])C(F)(F)F
C8H3F6NO2
MFCD00236674
2,5-bis trifluoromethyl nitrobenzene, 2-nitro-1,4-bis trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-nitrobenzene, 1-nitro-2,5-bis trifluoromethyl benzene, benzene, 2-nitro-1,4-bis trifluoromethyl, 2,5-bistrifluoromethylnitrobenzene, pubchem19738, 2-nitro-1,4-bis-trifluoromethyl-benzene, acmc-1bn6y, ksc496c8f
2-nitro-1,4-bis(trifluoromethyl)benzene

Specifications

320-88-7
C8H3F6NO2
5 g
BZKVUOHHNCMTLH-UHFFFAOYSA-N
2-nitro-1,4-bis(trifluoromethyl)benzene
2734048
≥98.0% (GC)
1-Nitro-2,5-bis(trifluoromethyl)benzene
90°C
MFCD00236674
2,5-bis trifluoromethyl nitrobenzene, 2-nitro-1,4-bis trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-nitrobenzene, 1-nitro-2,5-bis trifluoromethyl benzene, benzene, 2-nitro-1,4-bis trifluoromethyl, 2,5-bistrifluoromethylnitrobenzene, pubchem19738, 2-nitro-1,4-bis-trifluoromethyl-benzene, acmc-1bn6y, ksc496c8f
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])C(F)(F)F
259.107
259.11
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'