missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Nitro-4-n-propylbenzene, 96%, Thermo Scientific Chemicals

$936.84 - $936.84

Chemical Identifiers

CAS 10342-59-3
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00051839
InChI Key SXQBFCVVZIYXHV-UHFFFAOYSA-N
Synonym 1-nitro-4-propyl-benzene, 1-nitro-4-n-propylbenzene, 4-nitro-n-propylbenzene, 4-propylnitrobenzene, benzene, 1-nitro-4-propyl, p-nitropropylbenzene, p-propylnitrobenzene, 1-nitro-4-propylbenzol
PubChem CID 82559
IUPAC Name 1-nitro-4-propylbenzene
SMILES CCCC1=CC=C(C=C1)[N+]([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0773014
View Documents
Thermo Scientific Chemicals
L0773014
25 g N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10342-59-3
165.19
SXQBFCVVZIYXHV-UHFFFAOYSA-N
82559
CCCC1=CC=C(C=C1)[N+]([O-])=O
C9H11NO2
MFCD00051839
1-nitro-4-propyl-benzene, 1-nitro-4-n-propylbenzene, 4-nitro-n-propylbenzene, 4-propylnitrobenzene, benzene, 1-nitro-4-propyl, p-nitropropylbenzene, p-propylnitrobenzene, 1-nitro-4-propylbenzol
1-nitro-4-propylbenzene

Specifications

10342-59-3
96°C to 100°C (2 mmHg)
1.537
25 g
2043939
SXQBFCVVZIYXHV-UHFFFAOYSA-N
1-nitro-4-propylbenzene
82559
96%
-14°C
C9H11NO2
MFCD00051839
UN2810
1-nitro-4-propyl-benzene, 1-nitro-4-n-propylbenzene, 4-nitro-n-propylbenzene, 4-propylnitrobenzene, benzene, 1-nitro-4-propyl, p-nitropropylbenzene, p-propylnitrobenzene, 1-nitro-4-propylbenzol
CCCC1=CC=C(C=C1)[N+]([O-])=O
165.19
165.19
1-Nitro-4-n-propylbenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 233-743-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only