missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals 1-Nitroso-2-naphthol-3,6-disulfonic acid, disodium salt hydrate, pure, indicator gra

$65.98 - $676.65

Chemical Identifiers

CAS 525-05-3
Molecular Formula C10H5NNa2O8S2
Molecular Weight (g/mol) 377.25
MDL Number MFCD00149246
InChI Key DMKMTGULLYISBH-UHFFFAOYSA-L
Synonym nitroso-r salt, nitroso r, sodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate, disodium 1-nitroso-2-naphthol-3,6-disulfonate, nitroso r salt, 1-nitroso-3,6-disulfonate, 2,7-naphthalenedisulfonic acid, 3-hydroxy-4-nitroso-, disodium salt, 3-hydroxy-4-nitroso-2,7-naphthalenedisulfonic acid disodium salt, 1-nitroso-2-naphthol-3,6-disulfonic acid disodium salt, disodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulphonate
PubChem CID 10672
IUPAC Name disodium;3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate
SMILES [Na+].[Na+].OC1=C(N=O)C2=CC=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC346550250
SDS View Documents
Thermo Scientific Chemicals
346550250
25 g Glass bottle
Each for $65.98
 
AC346551000
SDS View Documents
Thermo Scientific Chemicals
346551000
100 g Glass bottle
Each for $156.81
 
AC346555000
SDS View Documents
Thermo Scientific Chemicals
346555000
500 g Glass bottle
Each for $676.65
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

525-05-3
377.25
DMKMTGULLYISBH-UHFFFAOYSA-L
10672
[Na+].[Na+].OC1=C(N=O)C2=CC=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O
C10H5NNa2O8S2
MFCD00149246
nitroso-r salt, nitroso r, sodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate, disodium 1-nitroso-2-naphthol-3,6-disulfonate, nitroso r salt, 1-nitroso-3,6-disulfonate, 2,7-naphthalenedisulfonic acid, 3-hydroxy-4-nitroso-, disodium salt, 3-hydroxy-4-nitroso-2,7-naphthalenedisulfonic acid disodium salt, 1-nitroso-2-naphthol-3,6-disulfonic acid disodium salt, disodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulphonate
disodium;3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate

Specifications

525-05-3
Yellow to Orange
Authentic
C10H5NNa2O8S2
25 g
15, 6731
Solubility in water: slightly soluble in water
[Na+].[Na+].OC1=C(N=O)C2=CC=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O
377.25
377.25
Crystalline Powder
>300.0°C
5% max. (1 g, 105°C)
Glass bottle
MFCD00149246
11, II, 190
nitroso-r salt, nitroso r, sodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate, disodium 1-nitroso-2-naphthol-3,6-disulfonate, nitroso r salt, 1-nitroso-3,6-disulfonate, 2,7-naphthalenedisulfonic acid, 3-hydroxy-4-nitroso-, disodium salt, 3-hydroxy-4-nitroso-2,7-naphthalenedisulfonic acid disodium salt, 1-nitroso-2-naphthol-3,6-disulfonic acid disodium salt, disodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulphonate
DMKMTGULLYISBH-UHFFFAOYSA-L
disodium;3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate
10672
Pure
1-Nitroso-2-naphthol-3, 6-disulfonic acid, disodium salt hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-369-1

RTECSNumber : QJ6480000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only