missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Nonanol, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 157470010

 View more versions of this product

Catalog No. AC157470010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Nonanol
96.0
-5.0°C
0.8270g/mL
74°C
96% min. (GC)
C9H20O
CH3(CH2)8OH
1 L
01, 423
15, 6766
Solubility in water: 1g/L (20°C)
CCCCCCCCCO
144.26
CHEBI:35986
97%
143-08-8
100.0
Colorless
213.0°C to 215.0°C
Authentic
Glass bottle
1.4324 to 1.4344
MFCD00002990
3082
0.827
1-nonanol, nonanol, n-nonyl alcohol, nonyl alcohol, pelargonic alcohol, octyl carbinol, 1-hydroxynonane, nonalol, n-nonanol, n-nonan-1-ol
ZWRUINPWMLAQRD-UHFFFAOYSA-N
nonan-1-ol
8914
144.26
Liquid

Chemical Identifiers

143-08-8
144.26
ZWRUINPWMLAQRD-UHFFFAOYSA-N
8914
nonan-1-ol
C9H20O
MFCD00002990
1-nonanol, nonanol, n-nonyl alcohol, nonyl alcohol, pelargonic alcohol, octyl carbinol, 1-hydroxynonane, nonalol, n-nonanol, n-nonan-1-ol
CHEBI:35986
CCCCCCCCCO
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 205-583-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only